accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSOOHO

CAS RN 5397-34-2

IUPAC Name: 4-(2-hydroxybenzenesulfonyl)phenol
Synonyms: 2,4'-sulfonyldiphenol
Molecular Weight (g/mol): 250.27
Molecular Formula: C12H10O4S
InChi Key: LROZSPADHSXFJA-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results