accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFFFH2N

CAS RN 53982-03-9

IUPAC Name: 3-methoxy-2-(trifluoromethyl)aniline
Synonyms: 3-methoxy-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: PBJBDVWRFMTPOI-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results