accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOOO

CAS RN 54-06-8

IUPAC Name: 3-hydroxy-1-methyl-2,3,5,6-tetrahydro-1H-indole-5,6-dione
Synonyms: adrenochrome
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: RPHLQSHHTJORHI-UHFFFAOYNA-N
SMILES: CN1CC(O)C2=CC(=O)C(=O)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Industry Type 
  • (3)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results