accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaCH3HgSOO

CAS RN 54-64-8

IUPAC Name: sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
Synonyms: thimerosal thiomersal ethylmercurithiosalicylate sodium 硫柳汞 sodium ethylmercurithiosalicylate thiomersalate mercurothiolate merthiolate
Molecular Weight (g/mol): 404.81
Molecular Formula: C9H9HgNaO2S
InChi Key: RTKIYNMVFMVABJ-UHFFFAOYSA-L
SMILES: [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O
Physical Form 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results