accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54-71-7

IUPAC Name: hydrogen (3S,4R)-3-ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-one chloride
Synonyms: hydrogen pilocarpine chloride
Molecular Weight (g/mol): 244.72
Molecular Formula: C11H17ClN2O2
InChi Key: RNAICSBVACLLGM-GNAZCLTHSA-N
SMILES: [H+].[Cl-].CC[C@H]1[C@@H](CC2=CN=CN2C)COC1=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results