accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2N

CAS RN 54-96-6

IUPAC Name: pyridine-3,4-diamine
Synonyms: 3,4-diaminopyridine amifampridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: OYTKINVCDFNREN-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results