accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 540-42-1

H3COOCH3CH3

CAS RN 540-42-1

IUPAC Name: 2-methylpropyl propanoate
Synonyms: isobutyl propionate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: FZXRXKLUIMKDEL-UHFFFAOYSA-N
SMILES: CCC(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results