accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3CH3H3C

CAS RN 540-88-5

IUPAC Name: tert-butyl acetate
Synonyms: tert-butyl acetate T-butyl acetate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: WMOVHXAZOJBABW-UHFFFAOYSA-N
SMILES: CC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)