accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3NSHOH3C

CAS RN 54016-70-5

IUPAC Name: 3-ethyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium bromide
Synonyms: 3-ethyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium bromide
Molecular Weight (g/mol): 252.17
Molecular Formula: C8H14BrNOS
InChi Key: BDQRQMLWZJQQKS-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1=CSC(CCO)=C1C
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results