accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOHO

CAS RN 54022-96-7

IUPAC Name: 5-(3-methoxyphenyl)furan-2-carboxylic acid
Synonyms: 5-(3-methoxyphenyl)furan-2-carboxylic acid
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: RIIFZAPFLSKUTL-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=CC=C(O1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results