accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 540475-45-4

FeP(R)(R)P(R)(R)CH3CH3CH3CH3

CAS RN 540475-45-4

IUPAC Name: λ²-iron(2+) bis(1-cyclopentyl-2,5-dimethylphospholane) dimethanide
Synonyms: λ²-iron(2+) bis(1-cyclopentyl-2,5-dimethylphospholane) dimethanide
Molecular Weight (g/mol): 454.44
Molecular Formula: C24H48FeP2
InChi Key: JBWUTRKMUDYCHS-UHFFFAOYNA-N
SMILES: [CH3-].[CH3-].[Fe++].CC1CCC(C)P1C1CCCC1.CC1CCC(C)P1C1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results