accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOH2N

CAS RN 540753-13-7

IUPAC Name: 1-{2-[2-(morpholin-4-yl)ethoxy]phenyl}methanamine
Synonyms: 1-{2-[2-(morpholin-4-yl)ethoxy]phenyl}methanamine
Molecular Weight (g/mol): 236.32
Molecular Formula: C13H20N2O2
InChi Key: QHAMTJRGPZWTSO-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1OCCN1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results