accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiH3COSiCH3CH3OSiH3CH3CO

CAS RN 541-05-9

IUPAC Name: hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Synonyms: cyclomethicone hexamethylcyclotrisiloxane
Molecular Weight (g/mol): 222.46
Molecular Formula: C6H18O3Si3
InChi Key: HTDJPCNNEPUOOQ-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results