accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 541-22-0

BrBrH3CNCH3H3CNCH3CH3H3C

CAS RN 541-22-0

IUPAC Name: trimethyl[10-(trimethylazaniumyl)decyl]azanium dibromide
Synonyms: decamethonium bromide
Molecular Weight (g/mol): 418.3
Molecular Formula: C16H38Br2N2
InChi Key: HLXQFVXURMXRPU-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results