accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 541-28-6

CH3CH3I

CAS RN 541-28-6

IUPAC Name: 1-iodo-3-methylbutane
Synonyms: butane, 1-iodo-3-methyl-
Molecular Weight (g/mol): 198.05
Molecular Formula: C5H11I
InChi Key: BUZZUHJODKQYTF-UHFFFAOYSA-N
SMILES: CC(C)CCI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)