accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 541-31-1

IUPAC Name: 3-methylbutane-1-thiol
Synonyms: isoamylthiol
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: GIJGXNFNUUFEGH-UHFFFAOYSA-N
SMILES: CC(C)CCS
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)