accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SNH3C

CAS RN 541-58-2

IUPAC Name: 2,4-dimethyl-1,3-thiazole
Synonyms: 2,4-dimethylthiazole
Molecular Weight (g/mol): 113.18
Molecular Formula: C5H7NS
InChi Key: OBSLLHNATPQFMJ-UHFFFAOYSA-N
SMILES: CC1=NC(C)=CS1
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)