accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5411-14-3

OHOOOHOO

CAS RN 5411-14-3

IUPAC Name: 2-[2-(carboxymethoxy)phenoxy]acetic acid
Synonyms: 2-(carboxymethoxy)phenoxyacetic acid
Molecular Weight (g/mol): 226.18
Molecular Formula: C10H10O6
InChi Key: PPZYHOQWRAUWAY-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1OCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results