accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHBr

CAS RN 5411-56-3

IUPAC Name: 1-(2-bromophenyl)ethan-1-ol
Synonyms: 1-(2-bromophenyl)ethanol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: DZLZSFZSPIUINR-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results