accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5416-93-3

IUPAC Name: 1-isocyanato-4-methoxybenzene
Synonyms: 1-isocyanato-4-methoxybenzene
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: FMDGXCSMDZMDHZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)