accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 542-28-9

IUPAC Name: oxan-2-one
Synonyms: delta-valerolactone delta-valeryllactone penta-1,5-lactone 5-valerolactone
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: OZJPLYNZGCXSJM-UHFFFAOYSA-N
SMILES: O=C1CCCCO1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)