accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 542-56-3

IUPAC Name: 2-methylpropyl nitrite
Synonyms: isobutyl nitrite isobutylnitrit isobutyl nitrite (substance) IBN blackjack
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: APNSGVMLAYLYCT-UHFFFAOYSA-N
SMILES: CC(C)CON=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)