accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54231-33-3

IUPAC Name: 3-bromo-2-nitropyridine
Synonyms: 3-bromo-2-nitropyridine
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: WFNISJZUJCKTLT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=NC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results