accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 543-20-4

IUPAC Name: butanedioyl dichloride
Synonyms: succinyl chloride
Molecular Weight (g/mol): 154.97
Molecular Formula: C4H4Cl2O2
InChi Key: IRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILES: ClC(=O)CCC(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (4)