accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 543-59-9

H3CCl

CAS RN 543-59-9

IUPAC Name: 1-chloropentane
Synonyms: 1-chloropentane
Molecular Weight (g/mol): 106.59
Molecular Formula: C5H11Cl
InChi Key: SQCZQTSHSZLZIQ-UHFFFAOYSA-N
SMILES: CCCCCCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)