accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BaCH3OOCH3OO

CAS RN 543-80-6

IUPAC Name: barium(2+) diacetate
Synonyms: barium acetate
Molecular Weight (g/mol): 255.42
Molecular Formula: C4H6BaO4
InChi Key: ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES: [Ba++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (18)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (9)
  • (1)
  • (5)
  • (3)
Grade 
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (7)
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (6)
  • (7)
  • (3)
  • (3)
  • (18)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (9)
  • (1)
  • (5)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)