accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OBr

CAS RN 54314-84-0

IUPAC Name: [(3-bromopropoxy)methyl]benzene
Synonyms: [(3-bromopropoxy)methyl]benzene
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: PSUXTZLDBVEZTD-UHFFFAOYSA-N
SMILES: BrCCCOCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results