CAS RN 5432-85-9
CAS RN 5432-85-9
IUPAC Name:
4-(propan-2-yl)cyclohexan-1-one
Synonyms:
4-isopropylcyclohexan-1-one
Molecular Weight (g/mol):
140.23
Molecular Formula:
C9H16O
InChi Key:
FPKISACHVIIMRA-UHFFFAOYSA-N
SMILES:
CC(C)C1CCC(=O)CC1
1
–
1
of
1
results
1
4-Isopropylcyclohexanone, 96%
CAS: 5432-85-9 Molecular Formula: C9H16O Molecular Weight (g/mol): 140.23 MDL Number: MFCD00043479 InChI Key: FPKISACHVIIMRA-UHFFFAOYSA-N Synonym: 4-isopropylcyclohexanone,cyclohexanone, 4-1-methylethyl,4-propan-2-yl cyclohexan-1-one,4-isopropylcyclohexan-1-one,cyclohexanone, 4-isopropyl,4-isopropyl-cyclohexanone,4-iso-propylcyclohexanone,4-methylethyl cyclohexan-1-one,acmc-20a5fk,4-isopropyl cyclohexanone PubChem CID: 79488 IUPAC Name: 4-propan-2-ylcyclohexan-1-one SMILES: CC(C)C1CCC(=O)CC1