accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3OH3CH3C

CAS RN 54344-92-2

IUPAC Name: 4-methoxy-2,3,6-trimethylbenzaldehyde
Synonyms: 4-methoxy-2,3,6-trimethylbenzaldehyde
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: BTOFIDLWQJCUJG-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C=O)C(C)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results