accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5437-38-7

CH3OHONOO

CAS RN 5437-38-7

IUPAC Name: 3-methyl-2-nitrobenzoic acid
Synonyms: 3-methyl-2-nitrobenzoic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: DGDAVTPQCQXLGU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results