accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONO

CAS RN 544-16-1

IUPAC Name: butyl nitrite
Synonyms: butyl nitrite
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: JQJPBYFTQAANLE-UHFFFAOYSA-N
SMILES: CCCCON=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results