accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CaOOOO

CAS RN 544-17-2

IUPAC Name: calcium diformate
Synonyms: calcium formate
Molecular Weight (g/mol): 130.11
Molecular Formula: C2H2CaO4
InChi Key: CBOCVOKPQGJKKJ-UHFFFAOYSA-L
SMILES: [Ca++].[O-]C=O.[O-]C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results