CAS RN 544-31-0
IUPAC Name:
N-(2-hydroxyethyl)hexadecanamide
Synonyms:
palmitoylethanolamide palmitoyl-EA anandamide (16:0) palmitamide mea palmitoyl ethanolamide
Molecular Weight (g/mol):
299.5
Molecular Formula:
C18H37NO2
InChi Key:
HXYVTAGFYLMHSO-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)NCCO
1
–
2
of
2
results
1
Palmitoylethanolamide, TRC
CAS: 544-31-0 Molecular Formula: C18H37NO2 Molecular Weight (g/mol): 299.49 Synonym: (Hydroxyethyl)palmitamide,2-(Palmitoylamino)ethanol,2-Palmitamidoethanol,AM 3112,Impulsin,Loramine P 256,Mackpeart DR 14V,N-(2-Hydroxyethyl)hexadecanamide,N-(2-Hydroxyethyl)palmitamide,N-Hexadecanoylethanolamine,N-Palmitoylethanolamine,NSC 23320,Palmidrol,Palmitic Acid Monoethanolamide,Palmitic Monoethanolamide IUPAC Name: N-(2-hydroxyethyl)hexadecanamide SMILES: CCCCCCCCCCCCCCCC(=O)NCCO
2
R&D Systems™ Palmitoylethanolamide
CAS: 544-31-0 Molecular Formula: C18H37NO2