CAS RN 544-35-4
IUPAC Name:
ethyl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms:
ethyl linoleate ethyl linoleate, pure pure ethyl linoleate mandenol
Molecular Weight (g/mol):
308.51
Molecular Formula:
C20H36O2
InChi Key:
FMMOOAYVCKXGMF-MURFETPASA-N
SMILES:
CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC
1
–
1
of
1
results
1
Ethyl Linoleate, TRC
CAS: 544-35-4 Molecular Formula: C20 H36 O2 Molecular Weight (g/mol): 308.5 Synonym: Ethyl Linoleate,(9Z,12Z)-Octadeca-9,12-dienoic Acid Ethyl Ester IUPAC Name: ethyl (9Z,12Z)-octadeca-9,12-dienoate SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC