accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3

CAS RN 544-40-1

IUPAC Name: 1-(butylsulfanyl)butane
Synonyms: dibutyl sulphide
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: HTIRHQRTDBPHNZ-UHFFFAOYSA-N
SMILES: CCCCSCCCC
Element 
  • (9)
Industry Type 
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Solvent or Matrix 
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results