accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 544-63-8

H3COHO

CAS RN 544-63-8

IUPAC Name: tetradecanoic acid
Synonyms: myristic acid acide tetradecanoique hydrofol acid 1495 myristic acid pure 1-tetradecanecarboxylic acid myristic acid, pure myristic acid (natural) hystrene 9014 acid, myristic pure myristic acid
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: TUNFSRHWOTWDNC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(O)=O
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)