accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CI

CAS RN 544-77-4

IUPAC Name: 1-iodohexadecane
Synonyms: 1-iodohexadecane
Molecular Weight (g/mol): 352.34
Molecular Formula: C16H33I
InChi Key: KMWHQYDMBYABKL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCI
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)