accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CuCN

CAS RN 544-92-3

IUPAC Name: λ¹-copper(1+) iminomethanide
Synonyms: copper(I) cyanide
Molecular Weight (g/mol): 89.56
Molecular Formula: CCuN
InChi Key: DOBRDRYODQBAMW-UHFFFAOYSA-N
SMILES: [Cu+].[C-]#N
Molecular Weight (g/mol) 
  • (6)
  • (9)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (6)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (9)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (9)
  • (3)