CAS RN 5443-49-2
IUPAC Name:
(2E)-2-bromo-3-phenylprop-2-enal
Synonyms:
bromocinnamal
Molecular Weight (g/mol):
211.06
Molecular Formula:
C9H7BrO
InChi Key:
WQRWNOKNRHCLHV-RMKNXTFCSA-N
SMILES:
Br\C(C=O)=C\C1=CC=CC=C1
1
–
2
of
2
results