accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 54443-89-9

CH3NOONCH3S

CAS RN 54443-89-9

IUPAC Name: 1,3-dibutyl-2-sulfanylidene-1,3-diazinane-4,6-dione
Synonyms: 1,3-dibutyl-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Weight (g/mol): 256.36
Molecular Formula: C12H20N2O2S
InChi Key: JNFBSDIDVPDZHB-UHFFFAOYSA-N
SMILES: CCCCN1C(=O)CC(=O)N(CCCC)C1=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results