CAS RN 5447-97-2
IUPAC Name:
2-bromo-2-nitropropane
Synonyms:
2-bromo-2-nitro-propane
Molecular Weight (g/mol):
167.99
Molecular Formula:
C3H6BrNO2
InChi Key:
OADSZWXMXIWZSQ-UHFFFAOYSA-N
SMILES:
CC(C)(Br)[N+]([O-])=O
1
–
1
of
1
results
1
2-Bromo-2-nitropropane, 98%
CAS: 5447-97-2 Molecular Formula: C3H6BrNO2 Molecular Weight (g/mol): 167.99 MDL Number: MFCD00007389 InChI Key: OADSZWXMXIWZSQ-UHFFFAOYSA-N Synonym: propane, 2-bromo-2-nitro,2-bromo-2-nitro-propane,snxhevlqiijip@,acmc-1aqp4,propane,2-bromo-2-nitro,2-bromo-2-nitropropane,oadszwxmxiwzsq-uhfffaoysa PubChem CID: 79531 IUPAC Name: 2-bromo-2-nitropropane SMILES: CC(C)([N+](=O)[O-])Br