CAS RN 545-46-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Uvaol, TRC
CAS: 545-46-0 Molecular Formula: C30 H50 O2 Molecular Weight (g/mol): 442.72 Synonym: (3β)-Urs-12-ene-3,28-diol,3,28-Dixydroxyurs-12-ene,3β,28-Dihydroxyurs-12-ene,Uvalol IUPAC Name: (3S,4aR,6aR,6bS,8aS,11R,12S,12aS,14aR,14bR)-8a-(hydroxymethyl)-4,4,6a,6b,11,12,14b-heptamethyl-2,3,4a,5,6,7,8,9,10,11,12,12a,14,14a-tetradecahydro-1H-picen-3-ol SMILES: C[C@@H]1CC[C@]2(CO)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C