accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2C(R)(R)H3CCH3(R)(R)(R)H3C(S)OHCH3H3C(R)(R)CH3(R)

CAS RN 545-47-1

IUPAC Name: (1R,3aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a,5a,5b,8,8,11a-hexamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
Synonyms: lupeol
Molecular Weight (g/mol): 426.73
Molecular Formula: C30H50O
InChi Key: MQYXUWHLBZFQQO-JGGBMTAGSA-N
SMILES: CC(=C)[C@@H]1CC[C@]2(C)CCC3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results