accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONOOH3C

CAS RN 5453-67-8

IUPAC Name: 2,6-dimethyl pyridine-2,6-dicarboxylate
Synonyms: 2,6-dimethyl pyridine-2,6-dicarboxylate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: SNQQJEJPJMXYTR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=N1)C(=O)OC
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results