accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOClCl

CAS RN 54551-83-6

IUPAC Name: methyl 2-(2,6-dichlorophenyl)acetate
Synonyms: methyl 2-(2,6-dichlorophenyl)acetate
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: FCWRUYPZZJPCCG-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results