accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5459-93-8

IUPAC Name: N-ethylcyclohexanamine
Synonyms: cyclohexanamine, N-ethyl-
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: AGVKXDPPPSLISR-UHFFFAOYSA-N
SMILES: CCNC1CCCCC1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results