accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 546-43-0

IUPAC Name: (3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-2H,3H,3aH,5H,6H,7H,8H,8aH,9H,9aH-naphtho[2,3-b]furan-2-one
Synonyms: helenine eupatal alant camphor alantolactone (substance) elecampane camphor alantolactone helenin inula camphor
Molecular Weight (g/mol): 232.32
Molecular Formula: C15H20O2
InChi Key: PXOYOCNNSUAQNS-AGNJHWRGSA-N
SMILES: C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12
Quantity 
  • (2)
  • (1)
  • (2)
Product Line 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results