accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PbCH3OOCH3OOCH3OOCH3OO

CAS RN 546-67-8

IUPAC Name: λ²-lead(2+) tetraacetate
Synonyms: lead(IV) acetate
Molecular Weight (g/mol): 443.4
Molecular Formula: C8H12O8Pb
InChi Key: ACKFDYCQCBEDNU-UHFFFAOYSA-J
SMILES: [Pb++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
  • (8)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (6)
  • (4)
Grade 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (6)
  • (4)
  • (3)
  • (2)
  • (3)