accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 546-68-9

IUPAC Name: titanium(4+) tetrakis(propan-2-olate)
Synonyms: titanium isopropoxide
Molecular Weight (g/mol): 284.22
Molecular Formula: C12H28O4Ti
InChi Key: VXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES: [Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Molecular Weight (g/mol) 
  • (10)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (3)
  • (4)
  • (3)
Physical Form 
  • (11)
Boiling Point 
  • (6)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (10)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (3)
  • (11)
  • (6)
  • (4)