accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaCH3OSOOO

CAS RN 546-74-7

IUPAC Name: sodium ethyl sulfate
Synonyms: sodium ethyl sulfate
Molecular Weight (g/mol): 148.11
Molecular Formula: C2H5NaO4S
InChi Key: RRLOOYQHUHGIRJ-UHFFFAOYSA-M
SMILES: [Na+].CCOS([O-])(=O)=O
Industry Type 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results